CAS RN 7440-69-9
Bismuth powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N Synonym: bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]
Bismuth, 99%, powder, -100 mesh, Thermo Scientific Chemicals
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N Synonym: bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]
Bismuth rod, 12.7mm (0.5in) dia, 99.999+% (metals basis), Thermo Scientific Chemicals
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N Synonym: bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]
Bismuth powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N Synonym: bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]
Bismuth foil, 2.0mm (0.08 in.) thick, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N Synonym: bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]
Bismuth powder, -200 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N Synonym: bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]
Bismuth, plasma standard solution, Specpure™ Bi 1000μg/mL
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]
Bismuth shot, 1-5mm (0.04-0.20in), 99.999% (metal basis), Thermo Scientific Chemicals
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N Synonym: bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]
Bismuth polycrystalline lump, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N Synonym: bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]
Bismuth foil, 1.0mm (0.04in) thick, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N Synonym: bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]
Bismuth granules, 1-2mm (0.04-0.08in), 99.997% (metals basis), Thermo Scientific Chemicals
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N Synonym: bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]
Bismuth needles, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N Synonym: bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]
Bismuth needles, 1-5cm (0.4-2in) long, 99.998% (metals basis), Thermo Scientific Chemicals
CAS: 7440-69-9 Molecular Formula: Bi Molecular Weight (g/mol): 208.98 MDL Number: MFCD00134033 InChI Key: JCXGWMGPZLAOME-UHFFFAOYSA-N Synonym: bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod PubChem CID: 5359367 ChEBI: CHEBI:33301 IUPAC Name: bismuth SMILES: [Bi]