accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OTeO

CAS RN 7446-07-3

IUPAC Name: (oxo-λ⁴-tellanylidene)oxidane
Synonyms: tellurite (teO2)
Molecular Weight (g/mol): 159.6
Molecular Formula: O2Te
InChi Key: LAJZODKXOMJMPK-UHFFFAOYSA-N
SMILES: O=[Te]=O
Molecular Weight (g/mol) 
  • (3)
  • (11)
Physical Form 
  • (12)
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
Grade 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (10)
  • (3)
  • (11)
  • (12)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)