accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7446-18-6

TlTlOSOOO

CAS RN 7446-18-6

IUPAC Name: bis(λ¹-thallanylium) sulfate
Synonyms: thallium (I) sulfate thallium(I) sulfate
Molecular Weight (g/mol): 504.82
Molecular Formula: O4STl2
InChi Key: YTQVHRVITVLIRD-UHFFFAOYSA-L
SMILES: [Tl+].[Tl+].[O-]S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (7)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (4)
  • (3)
  • (1)
Grade 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (7)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)