accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OZnOSOOO

CAS RN 7446-19-7

IUPAC Name: zinc(2+) hydrate sulfate
Synonyms: zinc(2+) hydrate sulfate
Molecular Weight (g/mol): 179.45
Molecular Formula: H2O5SZn
InChi Key: RNZCSKGULNFAMC-UHFFFAOYSA-L
SMILES: O.[Zn++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)