accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7447-39-4

ClClCu

CAS RN 7447-39-4

IUPAC Name: copper(2+) dichloride
Synonyms: cupric chloride
Molecular Weight (g/mol): 134.45
Molecular Formula: Cl2Cu
InChi Key: ORTQZVOHEJQUHG-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Cu++]
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (6)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (6)
  • (3)