accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 745055-86-1

NONOOOHH2NOOH

CAS RN 745055-86-1

IUPAC Name: 2-{[3-(2-amino-2-carboxyethyl)-2,6-dioxo-1,2,3,6-tetrahydropyrimidin-1-yl]methyl}benzoic acid
Synonyms: 2-{[3-(2-amino-2-carboxyethyl)-2,6-dioxopyrimidin-1-yl]methyl}benzoic acid
Molecular Weight (g/mol): 333.3
Molecular Formula: C15H15N3O6
InChi Key: UUIYULWYHDSXHL-UHFFFAOYNA-N
SMILES: NC(CN1C=CC(=O)N(CC2=CC=CC=C2C(O)=O)C1=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results