accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 74563-64-7

IUPAC Name: 3,7,11,15-tetramethylhexadecane-1,2,3-triol
Synonyms: phytantriol
Molecular Weight (g/mol): 330.55
Molecular Formula: C20H42O3
InChi Key: CGIHFIDULQUVJG-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)CCCC(C)CCCC(C)(O)C(O)CO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)