accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOFFF

CAS RN 745783-91-9

IUPAC Name: 2-oxo-2-[2-(trifluoromethyl)phenyl]acetaldehyde hydrate
Synonyms: 2-oxo-2-[2-(trifluoromethyl)phenyl]acetaldehyde hydrate
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: HBDWKZYNXMESGA-UHFFFAOYSA-N
SMILES: O.FC(F)(F)C1=CC=CC=C1C(=O)C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results