CAS RN 74663-48-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-Isopropyl-6-propylphenol, TRC
CAS: 74663-48-2 Molecular Formula: C12 H18 O Molecular Weight (g/mol): 178.27 Synonym: 2-(1-Methylethyl)-6-propylphenol IUPAC Name: 2-propan-2-yl-6-propylphenol SMILES: CCCc1cccc(C(C)C)c1O
2
2-(1-Methylethyl)-6-propylphenol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.