accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3OHOOH3C

CAS RN 74737-05-6

IUPAC Name: 3-(2,4,6-trimethoxyphenyl)propanoic acid
Synonyms: 3-(2,4,6-trimethoxyphenyl)propanoic acid
Molecular Weight (g/mol): 240.26
Molecular Formula: C12H16O5
InChi Key: LKDSCRCVTKEXRX-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(CCC(O)=O)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results