accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7484-37-9

BrP

CAS RN 7484-37-9

IUPAC Name: triphenyl(3-phenylpropyl)phosphanium bromide
Synonyms: triphenyl(3-phenylpropyl)phosphanium bromide
Molecular Weight (g/mol): 461.38
Molecular Formula: C27H26BrP
InChi Key: RPUZOJFXAPSSJD-UHFFFAOYSA-M
SMILES: [Br-].C(CC1=CC=CC=C1)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results