accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7488-56-4

SSeS

CAS RN 7488-56-4

IUPAC Name: thione
Synonyms: selenium disulfide
Molecular Weight (g/mol): 143.09
Molecular Formula: S2Se
InChi Key: JNMWHTHYDQTDQZ-UHFFFAOYSA-N
SMILES: S=[Se]=S
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)