accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 749849-14-7

HOONBrN

CAS RN 749849-14-7

IUPAC Name: 6-bromoimidazo[1,2-a]pyridine-2-carboxylic acid
Synonyms: 6-bromoimidazo[1,2-a]pyridine-2-carboxylic acid
Molecular Weight (g/mol): 241.04
Molecular Formula: C8H5BrN2O2
InChi Key: FQBFPVLZLNEQOE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN2C=C(Br)C=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results