accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 75-03-6

IUPAC Name: iodoethane
Synonyms: ethyl iodide
Molecular Weight (g/mol): 155.97
Molecular Formula: C2H5I
InChi Key: HVTICUPFWKNHNG-UHFFFAOYSA-N
SMILES: CCI
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (4)
  • (3)