accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

II

CAS RN 75-11-6

IUPAC Name: diiodomethane
Synonyms: diiodomethane methylene iodide
Molecular Weight (g/mol): 267.84
Molecular Formula: CH2I2
InChi Key: NZZFYRREKKOMAT-UHFFFAOYSA-N
SMILES: ICI
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (6)
  • (1)
  • (4)
Physical Form 
  • (11)
Percent Purity 
  • (6)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (6)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (4)
  • (11)
  • (6)
  • (4)