CAS RN 75-12-7
Formamide, 99.5+%, extra pure
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88 PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
Formamide, 99%, for molecular biology, DNAse, RNAse and Protease free
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88 PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
Formamide, 99.5%, for analysis
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88 PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
Formamide (≥99.5%), Fisher BioReagents
Formamide, Certified AR for Analysis, Fisher Chemical™
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88 PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
Formamide, 99.6%, ACS reagent
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88 PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
Formamide, 99%
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88 PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
Formamide, 99+%, for spectroscopy
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88 PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
Formamide, Spectrophotometric Grade, 99+%
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88 PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
Formamide, Ultra Pure, 99.5% min
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88 PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
Formamide (Molecular Biology), Fisher BioReagents
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88 PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
Formamide, ACS, 99.5+%
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88 PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
HYDRANAL™ - Formamide dry, Solvent for KF titration, Solstice
Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
Formamide, puriss. p.a., ACS Reagent, ≥99.5% (GC/T), Solstice
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.04 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88 PubChem CID: 713 ChEBI: CHEBI:48431 IUPAC Name: formamide SMILES: NC=O
Formamide, TRC
CAS: 75-12-7 Molecular Formula: C H3 N O Molecular Weight (g/mol): 45.04 Synonym: Formamide,Carbamaldehyde,Formimidic acid,Methanamide,NSC 748 IUPAC Name: formamide SMILES: NC=O
- 1
- 2