accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 75-16-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 119.24
Molecular Formula: CH3BrMg
InChi Key: NXPHGHWWQRMDIA-UHFFFAOYSA-M
SMILES: [CH3-].[Mg++].[Br-]
Quantity 
  • (5)
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results