accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 75-18-3

IUPAC Name: (methylsulfanyl)methane
Synonyms: dimethyl sulfide dimethyl sulfide [UN1164] [flammable liquid] methylthiomethyl radical exact-S reduced-dmso methane, 1,1'-thiobis- 1,1'-thiobis-methane dimethyl sulfide (natural) sulfure de methyle dimethylsulfid
Molecular Weight (g/mol): 62.13
Molecular Formula: C2H6S
InChi Key: QMMFVYPAHWMCMS-UHFFFAOYSA-N
SMILES: CSC
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results