accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CaCC

CAS RN 75-20-7

IUPAC Name: calcium acetylene
Synonyms: calcium carbide
Molecular Weight (g/mol): 64.1
Molecular Formula: C2Ca
InChi Key: UIXRSLJINYRGFQ-UHFFFAOYSA-N
SMILES: [Ca++].[C-]#[C-]
Physical Form 
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)