CAS RN 75-29-6
IUPAC Name:
2-chloropropane
Synonyms:
2-chloropropane
Molecular Weight (g/mol):
78.54
Molecular Formula:
C3H7Cl
InChi Key:
ULYZAYCEDJDHCC-UHFFFAOYSA-N
SMILES:
CC(C)Cl
1
–
4
of
4
results
1
2-Chloropropane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Chloropropane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Chloropropane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.