accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SiClCl

CAS RN 75-54-7

IUPAC Name: dichloro(methyl)silyl
Synonyms: dichloro(methyl)silyl
Molecular Weight (g/mol): 114.02
Molecular Formula: CH3Cl2Si
InChi Key: KTQYJQFGNYHXMB-UHFFFAOYSA-N
SMILES: C[Si](Cl)Cl
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)