accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHCH3NCH3CH3H3C

CAS RN 75-59-2

IUPAC Name: tetramethylazanium hydroxide
Synonyms: tetramethylammonium hydroxide
Molecular Weight (g/mol): 91.15
Molecular Formula: C4H13NO
InChi Key: WGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES: [OH-].C[N+](C)(C)C
Molecular Weight (g/mol) 
  • (12)
  • (20)
Quantity 
  • (5)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (16)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (6)
Boiling Point 
  • (5)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (9)
  • (14)
  • (3)
  • (12)
  • (20)
  • (5)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (3)
  • (16)
  • (3)
  • (3)
  • (3)
  • (6)
  • (5)
  • (3)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)