accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOHOO

CAS RN 75-75-2

IUPAC Name: methanesulfonic acid
Synonyms: methanesulfonic acid mesylates kyselina methansulfonova iron (3+)salt methanesulfonic acid methanesulfonic acid, silver (1+) salt methanesulfonic acid, iron (3+)salt nickel (2+) salt methanesulfonic acid silver (1+) salt methanesulfonic acid MSA sodium salt methanesulfonic acid
Molecular Weight (g/mol): 96.1
Molecular Formula: CH4O3S
InChi Key: AFVFQIVMOAPDHO-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O
Molecular Weight (g/mol) 
  • (21)
  • (4)
Quantity 
  • (4)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (2)
  • (4)
Physical Form 
  • (11)
  • (3)
Boiling Point 
  • (3)
  • (1)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (9)
  • (6)
  • (3)
  • (1)
  • (21)
  • (4)
  • (4)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (4)
  • (11)
  • (3)
  • (3)
  • (1)
  • (6)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)