accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 75-77-4

IUPAC Name: chlorotrimethylsilane
Synonyms: chlorotrimethylsilane
Molecular Weight (g/mol): 108.64
Molecular Formula: C3H9ClSi
InChi Key: IJOOHPMOJXWVHK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)Cl
Quantity 
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (4)
  • (6)
Boiling Point 
  • (1)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (6)
  • (1)
  • (6)
  • (7)