accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH3CH3H3C

CAS RN 75-97-8

IUPAC Name: 3,3-dimethylbutan-2-one
Synonyms: pinacolone
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: PJGSXYOJTGTZAV-UHFFFAOYSA-N
SMILES: CC(=O)C(C)(C)C
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)