accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7512-17-6

IUPAC Name: N-[2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Synonyms: chitin
Molecular Weight (g/mol): 221.21
Molecular Formula: C8H15NO6
InChi Key: OVRNDRQMDRJTHS-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)OC(CO)C(O)C1O
Quantity 
  • (1)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (6)
  • (3)
  • (1)
  • (3)
  • (1)