accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOONHNOOH3CHN

CAS RN 75184-71-3

IUPAC Name: methyl N-[6-(propane-1-sulfonyl)-1H-1,3-benzodiazol-2-yl]carbamate
Synonyms: methyl N-[5-(propane-1-sulfonyl)-3H-1,3-benzodiazol-2-yl]carbamate
Molecular Weight (g/mol): 297.33
Molecular Formula: C12H15N3O4S
InChi Key: CLSJYOLYMZNKJB-UHFFFAOYSA-N
SMILES: CCCS(=O)(=O)C1=CC=C2N=C(NC(=O)OC)NC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results