accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 75272-39-8

IUPAC Name: N-[(2R,3R)-1-benzyl-2-methylpyrrolidin-3-yl]-5-chloro-2-methoxy-4-(methylamino)benzamide
Synonyms: (+)-nemonapride (2R,3R)-nemonapride
Molecular Weight (g/mol): 387.91
Molecular Formula: C21H26ClN3O2
InChi Key: KRVOJOCLBAAKSJ-RDTXWAMCSA-N
SMILES: CNC1=CC(OC)=C(C=C1Cl)C(=O)N[C@@H]1CCN(CC2=CC=CC=C2)[C@@H]1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results