accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClHSnClHCH3H3C

CAS RN 753-73-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 221.7
Molecular Formula: C2H8Cl2Sn
InChi Key: QLQTYMCOTSXVHH-UHFFFAOYSA-N
SMILES: Cl.Cl.C[SnH2]C
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (5)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)