accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 75350-46-8

IUPAC Name: 1-{3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-5-yl}-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-{3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthen]-5-yl}pyrrole-2,5-dione
Molecular Weight (g/mol): 427.37
Molecular Formula: C24H13NO7
InChi Key: AYDAHOIUHVUJHQ-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=CC(=CC=C23)N2C(=O)C=CC2=O)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results