CAS RN 7553-56-2
Iodine, 0.1N (0.05M) standard solution
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: diiodine SMILES: II
Iodine Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: molecular iodine SMILES: II
Iodine, 99+%
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N IUPAC Name: diiodine SMILES: II
Iodine, 99.5+%, Extra Pure, SLR, Fisher Chemical™
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 SMILES: II
Iodine, 99+%, pure
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 SMILES: II
Iodine, Certified AR for Analysis, Fisher Chemical™
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 SMILES: II
Thermo Scientific Chemicals Iodine, 0.01N Standardized Solution
CAS: 7553-56-2 | I2 | 253.81 g/mol
Iodine, 99.5%, extra pure, resublimed
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 SMILES: II
Iodine, 99.8+%, ACS reagent
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 SMILES: II
Iodine, crystalline, 99.99+% (metals basis)
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 SMILES: II
Iodine, specified according to the requirements of Ph.Eur, BP, USP
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: diiodine SMILES: II
Iodine, resublimed crystals, ACS, 99.8%
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: molecular iodine SMILES: II
Iodine, 99.5+%, for analysis
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: diiodine SMILES: II
Iodine Solution 0.5M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 SMILES: II
Iodine, resublimed crystals, Puratronic™, 99.9985% (metals basis)
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.81 MDL Number: MFCD00011355 MFCD00164163 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode PubChem CID: 807 ChEBI: CHEBI:17606 SMILES: II