accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2BrBrH2N

CAS RN 75568-11-5

IUPAC Name: 3,5-dibromobenzene-1,2-diamine
Synonyms: 3,5-dibromobenzene-1,2-diamine
Molecular Weight (g/mol): 265.94
Molecular Formula: C6H6Br2N2
InChi Key: BABDSAMUOCWCNS-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=CC(Br)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results