accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OFFFFFFFFFFFF

CAS RN 756-13-8

IUPAC Name: 1,1,1,2,2,4,5,5,5-nonafluoro-4-(trifluoromethyl)pentan-3-one
Synonyms: 1,1,1,2,2,4,5,5,5-nonafluoro-4-(trifluoromethyl)pentan-3-one
Molecular Weight (g/mol): 316.05
Molecular Formula: C6F12O
InChi Key: RMLFHPWPTXWZNJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(=O)C(F)(C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results