accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 756-79-6

CH3OPCH3OOH3C

CAS RN 756-79-6

IUPAC Name: dimethyl methylphosphonate
Synonyms: dimethyl methylphosphonate
Molecular Weight (g/mol): 124.08
Molecular Formula: C3H9O3P
InChi Key: VONWDASPFIQPDY-UHFFFAOYSA-N
SMILES: COP(C)(=O)OC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results