accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3H3COOHN(R)H2NOOO

CAS RN 75647-01-7

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-carbamoylpropanoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-carbamoylpropanoate
Molecular Weight (g/mol): 231.23
Molecular Formula: C9H15N2O5
InChi Key: FYYSQDHBALBGHX-RXMQYKEDSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC(N)=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)