accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BHNSOOCH3HOOH

CAS RN 756520-78-2

IUPAC Name: (2-methanesulfonamidophenyl)boronic acid
Synonyms: 2-methanesulfonamidophenylboronic acid
Molecular Weight (g/mol): 215.03
Molecular Formula: C7H10BNO4S
InChi Key: RPRMOEFOTPSWBO-UHFFFAOYSA-N
SMILES: CS(=O)(=O)NC1=CC=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results