accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 75681-13-9

IUPAC Name: methyl 3-hydrazinylthiophene-2-carboxylate
Synonyms: methyl 3-hydrazinylthiophene-2-carboxylate
Molecular Weight (g/mol): 172.2
Molecular Formula: C6H8N2O2S
InChi Key: RWGYQWWIXLVGDJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(NN)C=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results