accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOHOHH2NHCl

CAS RN 75705-21-4

IUPAC Name: hydrogen [4-(aminomethyl)phenyl]boronic acid chloride
Synonyms: hydrogen 4-(aminomethyl)phenylboronic acid chloride
Molecular Weight (g/mol): 187.43
Molecular Formula: C7H11BClNO2
InChi Key: HUZNRXFJHYNUMV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC1=CC=C(C=C1)B(O)O
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)