CAS RN 757978-18-0
IUPAC Name:
5-bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine
Synonyms:
5-bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine
Molecular Weight (g/mol):
322.93
Molecular Formula:
C7H4BrIN2
InChi Key:
GIPGJYARDOQGDJ-UHFFFAOYSA-N
SMILES:
BrC1=CN=C2NC=C(I)C2=C1
1
–
1
of
1
results
1
5-Bromo-3-iodo-7-azaindole, 97%, Thermo Scientific™
CAS: 757978-18-0 Molecular Formula: C7H4BrIN2 Molecular Weight (g/mol): 322.931 MDL Number: MFCD07779519 InChI Key: GIPGJYARDOQGDJ-UHFFFAOYSA-N Synonym: 5-bromo-3-iodo-1h-pyrrolo 2,3-b pyridine,5-bromo-3-iodo-7-azaindole,1h-pyrrolo 2,3-b pyridine, 5-bromo-3-iodo,pubchem15707,endotherm enu336,ksc495s5j,5-bromo-3-iodopyrrolo 2,3-b pyridine,3-iodo-5-bromo-1h-pyrrolo 2,3-b-pyridine PubChem CID: 24229212 IUPAC Name: 5-bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine SMILES: C1=C2C(=CNC2=NC=C1Br)I