accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 758-08-7

NH2OHS

CAS RN 758-08-7

IUPAC Name: 2-sulfanylacetamide
Synonyms: sulfacetamide
Molecular Weight (g/mol): 91.13
Molecular Formula: C2H5NOS
InChi Key: GYXHHICIFZSKKZ-UHFFFAOYSA-N
SMILES: NC(=O)CS
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results