accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONCH3H3C

CAS RN 758-96-3

IUPAC Name: N,N-dimethylpropanamide
Synonyms: propanamide, N,N-dimethyl-
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: MBHINSULENHCMF-UHFFFAOYSA-N
SMILES: CCC(=O)N(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results