accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7580-67-8

LiH

CAS RN 7580-67-8

IUPAC Name: lithium(1+) hydride
Synonyms: lithium hydride
Molecular Weight (g/mol): 7.95
Molecular Formula: HLi
InChi Key: SRTHRWZAMDZJOS-UHFFFAOYSA-N
SMILES: [H-].[Li+]
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (4)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (6)
  • (3)
  • (4)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)