accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 75844-69-8

IUPAC Name: tert-butyl piperidine-1-carboxylate
Synonyms: tert-butyl piperidine-1-carboxylate
Molecular Weight (g/mol): 185.27
Molecular Formula: C10H19NO2
InChi Key: RQCNHUCCQJMSRG-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCCCC1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)