CAS RN 7585-39-9
beta-Cyclodextrin, 98%
CAS: 7585-39-9 Molecular Formula: C42H70O35 Molecular Weight (g/mol): 1134.99 MDL Number: MFCD00078139 InChI Key: WHGYBXFWUBPSRW-UHFFFAOYNA-N Synonym: cycloheptaamylose,cycloheptapentylose,.beta.-cyclodextrin,caraway,cyclo-hepta-amylose,cycloheptamaltose,.beta.-dextrin,beta-cycloamylose,beta-cyclodextrin,.beta.-cycloamylose PubChem CID: 131707246 IUPAC Name: 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol SMILES: OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
beta-Cyclodextrin
CAS: 7585-39-9 Molecular Formula: C42H70O35 Molecular Weight (g/mol): 1134.99 MDL Number: MFCD00078139 InChI Key: WHGYBXFWUBPSRW-UHFFFAOYNA-N Synonym: cycloheptaamylose,cycloheptapentylose,.beta.-cyclodextrin,caraway,cyclo-hepta-amylose,cycloheptamaltose,.beta.-dextrin,beta-cycloamylose,beta-cyclodextrin,.beta.-cycloamylose PubChem CID: 131707246 IUPAC Name: 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol SMILES: OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
β-Cyclodextrin, >98%, MP Biomedicals™
CAS: 7585-39-9 Molecular Formula: C42H70O35 Molecular Weight (g/mol): 1134.99 MDL Number: MFCD00078139 InChI Key: WHGYBXFWUBPSRW-UHFFFAOYNA-N Synonym: cycloheptaamylose,cycloheptapentylose,.beta.-cyclodextrin,caraway,cyclo-hepta-amylose,cycloheptamaltose,.beta.-dextrin,beta-cycloamylose,beta-cyclodextrin,.beta.-cycloamylose PubChem CID: 131707246 IUPAC Name: 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol SMILES: OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
Beta-Cyclodextrin, TRC
CAS: 7585-39-9 Molecular Formula: C42 H70 O35 Molecular Weight (g/mol): 1134.98 Synonym: Betadex,Cycloheptakis-(1→4)-(α-d-glucopyranosyl) (cyclomaltoheptaose),Cycloheptakis-(1→4)-(α-d-glucopyranosyl) β-cyclodextrin IUPAC Name: (1S,3R,5R,6S,8R,10R,11S,13R,15R,16S,18R,20R,21S,23R,25R,26S,28R,30R,31S,33R,35R,36R,37R,38R,39R,40R,41R,42R,43R,44R,45R,46R,47R,48R,49R)-5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.23,6.28,11.213,16.218,21.223,26.228,31]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol SMILES: OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]1[C@H](O)[C@H]2O
R&D Systems™ b-Cyclodextrin
A Tocris product, part of R&D Systems. Cyclic complexing agent