CAS RN 75885-58-4
IUPAC Name:
2,2-dimethylcyclopropane-1-carboxamide
Synonyms:
2,2-dimethylcyclopropane-1-carboxamide
Molecular Weight (g/mol):
113.16
Molecular Formula:
C6H11NO
InChi Key:
YBZQRYWKYBZZNT-UHFFFAOYNA-N
SMILES:
CC1(C)CC1C(N)=O
1
–
1
of
1
results
1
(1S)-2,2-Dimethylcyclopropanecarboxamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.