accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH3COSCH3

CAS RN 759-94-4

IUPAC Name: N,N-dipropyl(ethylsulfanyl)formamide
Synonyms: torbin caswell no. 435 genep eptc C9H19nos EPTC stauffer R 1608 niptan epa pesticide chemical code 041401 alirox eptc (herbicide)
Molecular Weight (g/mol): 189.32
Molecular Formula: C9H19NOS
InChi Key: GUVLYNGULCJVDO-UHFFFAOYSA-N
SMILES: CCCN(CCC)C(=O)SCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results