accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 75920-10-4

IUPAC Name: 1,4,8,11-tetraazatricyclo[9.3.1.1⁴,⁸]hexadecane
Synonyms: 1,4,8,11-tetraazatricyclo[9.3.1.1⁴,⁸]hexadecane
Molecular Weight (g/mol): 224.35
Molecular Formula: C12H24N4
InChi Key: ZJCWEHQOOSNWTF-UHFFFAOYNA-N
SMILES: C1CN2CN(C1)CCN1CCCN(C1)CC2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results