accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOClClCl

CAS RN 76-02-8

IUPAC Name: trichloroacetyl chloride
Synonyms: trichloroacetyl chloride
Molecular Weight (g/mol): 181.82
Molecular Formula: C2Cl4O
InChi Key: PVFOMCVHYWHZJE-UHFFFAOYSA-N
SMILES: ClC(=O)C(Cl)(Cl)Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results