accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClClClClClCl

CAS RN 76-44-8

IUPAC Name: 1,5,7,8,9,10,10-heptachlorotricyclo[5.2.1.0²,⁶]deca-3,8-diene
Synonyms: heptachlor heptachlor isomer 3-chlorochlordene heptachlorane heptamul
Molecular Weight (g/mol): 373.3
Molecular Formula: C10H5Cl7
InChi Key: FRCCEHPWNOQAEU-UHFFFAOYNA-N
SMILES: ClC1C=CC2C1C1(Cl)C(Cl)=C(Cl)C2(Cl)C1(Cl)Cl
Molecular Weight (g/mol) 
  • (4)
  • (3)
Solvent or Matrix 
  • (1)
  • (1)
  • (2)
Concentration 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)