accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HO

CAS RN 76-84-6

IUPAC Name: triphenylmethanol
Synonyms: triphenylmethanol
Molecular Weight (g/mol): 260.34
Molecular Formula: C19H16O
InChi Key: LZTRCELOJRDYMQ-UHFFFAOYSA-N
SMILES: OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)